2-Fluoro-3-methylpyridine-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

21994-1G 21994-500MG 21994-5G
76898-556EA 676.86 USD
76898-556 76898-558 76898-560
2-Fluoro-3-methylpyridine-4-carboxaldehyde ≥97%
2-Fluoro-3-methylpyridine-4-carboxaldehyde
Formula: C₇H₆FNO
MW: 139.127 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611269
CAS Number: 1211581-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR