2,6-Dibromo-3-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

21973-25G 21973-5G
76898-544EA 587.74 USD
76898-544 76898-546
2,6-Dibromo-3-methoxypyridine ≥97%
2,6-Dibromo-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MW: 266.918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26743441
CAS Number: 79491-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR