(6-Ethoxy-4-methylpyridin-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Ethoxy-4-methylpyridine-5-boronic acid

21925-1G 21925-25G 21925-500MG 21925-5G
76898-532EA 201.58 USD
76898-532 76898-534 76898-536 76898-538
(6-Ethoxy-4-methylpyridin-3-yl)boronic acid ≥97%
(6-Ethoxy-4-methylpyridin-3-yl)boronic acid
Formula: C₈H₁₂BNO₃
MW: 181.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374995
CAS Number: 1256355-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR