2-Bromo-5-chloronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

21807-25G 21807-5G
76898-480EA 559.1 USD
76898-480 76898-482
2-Bromo-5-chloronicotinic acid ≥97%
2-Bromo-5-chloronicotinic acid
Formula: C₆H₃BrClNO₂
Storage Temperature: Ambient
CAS Number: 65550-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR