2-Methoxy-5-(methoxymethoxy)-pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

21708-25G 21708-5G
76898-464EA 595.17 USD
76898-464 76898-466
2-Methoxy-5-(methoxymethoxy)-pyridine ≥97% research grade
2-Methoxy-5-(methoxymethoxy)-pyridine
Formula: C₈H₁₁NO₃
MW: 169.178 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26386947
CAS Number: 431942-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR