3,5-Difluoro-2-methoxyphenol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Difluoro-6-hydroxyanisole

21518-1G 21518-25G 21518-500MG 21518-5G
76898-422EA 159.14 USD
76898-422 76898-424 76898-426 76898-428
3,5-Difluoro-2-methoxyphenol ≥97% research grade
3,5-Difluoro-2-methoxyphenol
Formula: C₇H₆F₂O₂
MW: 160.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12404933
CAS Number: 152434-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR