2,5-Difluoro-4-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21252-1G 21252-25G 21252-500MG 21252-5G
76898-342EA 231.28 USD
76898-342 76898-344 76898-346 76898-348
2,5-Difluoro-4-hydroxyphenylboronic acid ≥97%
2,5-Difluoro-4-hydroxyphenylboronic acid
Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18394881
CAS Number: 1229584-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR