2-Bromo-5-formylthiazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromothiazole-5-carboxaldehyde

21177-25G 21177-5G
76898-318EA 268.41 USD
76898-318 76898-320
2-Bromo-5-formylthiazole ≥97%
2-Bromo-5-formylthiazole

Precaución: for Research Use Only

Formula: C₄H₂BrNOS
MW: 192.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03788567
CAS Number: 464192-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR