2,6-Difluoro-4-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21092-25G 21092-5G
76898-290EA 513.48 USD
76898-290 76898-292
2,6-Difluoro-4-hydroxybenzaldehyde ≥97%
2,6-Difluoro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07772042
CAS Number: 532967-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR