2,6-Dichloro-4-methyl-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

21087-1G 21087-25G 21087-5G
76898-282EA 62.6 USD
76898-282 76898-284 76898-286
2,6-Dichloro-4-methyl-benzoic acid ≥97%
2,6-Dichloro-4-methyl-benzoic acid
Formula: C₈H₆Cl₂O₂
MW: 205.038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447615
CAS Number: 99520-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR