Methyl-2-fluoro-6-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-fluoro-6-methoxybenzoate, 2-Fluoro-6-methoxybenzoic acid methyl ester

20974-25G 20974-5G
76898-256EA 310.85 USD
76898-256 76898-258
Methyl-2-fluoro-6-methoxybenzoate ≥97%
Methyl-2-fluoro-6-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.17
Storage Temperature: Ambient
MDL Number: MFCD03788537
CAS Number: 178747-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR