2,6-Dibromo-4-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

20939-25G 20939-5G
76898-238EA 453.01 USD
76898-238 76898-240
2,6-Dibromo-4-isopropylphenol ≥97%
2,6-Dibromo-4-isopropylphenol
Formula: C₉H₁₀Br₂O
Storage Temperature: Ambient
MDL Number: MFCD18447613
CAS Number: 2432-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR