2-Chloro-6-fluoro-3-methylanisole ≥97%

Supplier: AOBChem USA

Warning

20921-25G 20921-5G
76898-230EA 418 USD
76898-230 76898-232
2-Chloro-6-fluoro-3-methylanisole ≥97%
2-Chloro-6-fluoro-3-methylanisole

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MW: 174.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631367
CAS Number: 261762-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR