2-[4-Bromo-2-(trifluoromethyl)phenyl]-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

20859-1G 20859-25G 20859-5G
76898-210EA 116.7 USD
76898-210 76898-212 76898-214
2-[4-Bromo-2-(trifluoromethyl)phenyl]-1,3-dioxolane ≥95%
2-[4-Bromo-2-(trifluoromethyl)phenyl]-1,3-dioxolane
Formula: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619410
CAS Number: 2221812-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR