2,6-Difluoro-4-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

20841-25G 20841-5G
76898-186EA 381.93 USD
76898-186 76898-188
2,6-Difluoro-4-methoxybenzyl alcohol ≥97%
2,6-Difluoro-4-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115923
CAS Number: 79538-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR