2-Methoxy-5-methylnicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-5-methylpyridine-3-carboxaldehyde

20715-1G 20715-25G 20715-500MG 20715-5G
76898-158EA 393.6 USD
76898-158 76898-160 76898-162 76898-164
2-Methoxy-5-methylnicotinaldehyde ≥95%
2-Methoxy-5-methylnicotinaldehyde
Formula: C₈H₉NO₂
MW: 151.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13563060
CAS Number: 1203499-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR