2-Methoxy-5-methylbenzyl alcohol ≥97% research grade

Supplier: AOBChem USA

Warning

20703-1G 20703-25G 20703-5G
76898-148EA 95.49 USD
76898-148 76898-150 76898-152
2-Methoxy-5-methylbenzyl alcohol ≥97% research grade
2-Methoxy-5-methylbenzyl alcohol
Formula: C₉H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08442735
CAS Number: 7048-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR