5-Chloro-2-fluoronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

20696-25G 20696-5G
76898-136EA 502.87 USD
76898-136 76898-138
5-Chloro-2-fluoronicotinic acid ≥97%
5-Chloro-2-fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD04112518
CAS Number: 884494-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR