2-Fluoro-3-methoxypyridine-5-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

20641-1G 20641-25G 20641-500MG 20641-5G
76898-120EA 439.22 USD
76898-120 76898-122 76898-124 76898-126
2-Fluoro-3-methoxypyridine-5-carboxylic acid ≥97% research grade
2-Fluoro-3-methoxypyridine-5-carboxylic acid
Formula: C₇H₆FNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18257417
CAS Number: 1256819-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR