2-Formyl-4-thiopheneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Formylthiophene-4-boronic acid

20639-25G 20639-5G
76898-108EA 346.92 USD
76898-108 76898-110
2-Formyl-4-thiopheneboronic acid ≥97%
2-Formyl-4-thiopheneboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD03002365
CAS Number: 175592-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR