2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-5-(piperidine-1-carbonyl)phenyl)boronic acid, 2-Fluoro-5-(1-piperidinylcarbonyl)benzeneboronic acid

20591-1G 20591-25G 20591-500MG 20591-5G
76898-080EA 142.17 USD
76898-080 76898-082 76898-084 76898-086
2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid ≥97%
2-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₅BFNO₃
MW: 251.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08436068
CAS Number: 874289-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR