2-Bromo-5-(methoxymethyl)-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

20540-1G 20540-25G 20540-500MG 20540-5G
76898-030EA 173.99 USD
76898-030 76898-032 76898-034 76898-036
2-Bromo-5-(methoxymethyl)-1,3-dimethylbenzene ≥97%
2-Bromo-5-(methoxymethyl)-1,3-dimethylbenzene
Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088120
CAS Number: 74973-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR