2-Bromo-6-fluoro-3-methylanisole ≥97%

Supplier: AOBChem USA

Warning

20512-1G 20512-25G 20512-5G
76898-018EA 88.06 USD
76898-018 76898-020 76898-022
2-Bromo-6-fluoro-3-methylanisole ≥97%
2-Bromo-6-fluoro-3-methylanisole
Formula: C₈H₈BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD16295102
CAS Number: 1226808-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR