2-Bromo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20507-1G 20507-25G 20507-500MG 20507-5G
76898-004EA 152.77 USD
76898-004 76898-006 76898-008 76898-010
2-Bromo-3-methoxybenzaldehyde ≥97%
2-Bromo-3-methoxybenzaldehyde
Formula: C₈H₇BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09834663
CAS Number: 10401-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR