2-Fluoro-5-hydroxypyridine-4-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

20464-1G 20464-500MG 20464-5G
76897-988EA 706.56 USD
76897-988 76897-990 76897-992
2-Fluoro-5-hydroxypyridine-4-boronic acid ≥97% research grade
2-Fluoro-5-hydroxypyridine-4-boronic acid
Formula: C₅H₅BFNO₃
MW: 156.908 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611108
CAS Number: 2121511-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR