2-Fluoro-5-methoxynicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-methoxypyridine-3-carboxaldehyde

20456-1G 20456-25G 20456-500MG 20456-5G
76897-956EA 201.58 USD
76897-956 76897-958 76897-960 76897-962
2-Fluoro-5-methoxynicotinaldehyde ≥97%
2-Fluoro-5-methoxynicotinaldehyde
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611224
CAS Number: 1227597-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR