(2-Fluoro-4-hydroxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

20404-1G 20404-25G 20404-5G
76897-930EA 106.09 USD
76897-930 76897-932 76897-934
(2-Fluoro-4-hydroxyphenyl)boronic acid ≥97%
(2-Fluoro-4-hydroxyphenyl)boronic acid
Formula: C₆H₆BFO₃
Storage Temperature: Ambient
MDL Number: MFCD11101444
CAS Number: 1376989-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR