2-Fluoro-5-isopropoxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

20388-1G 20388-25G 20388-500MG 20388-5G
76897-918EA 171.87 USD
76897-918 76897-920 76897-922 76897-924
2-Fluoro-5-isopropoxypyridine ≥97% research grade
2-Fluoro-5-isopropoxypyridine
Formula: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD18802376
CAS Number: 904961-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR