2-Bromo-1,3,4-trichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

20374-1G 20374-25G 20374-5G
76897-910EA 62.6 USD
76897-910 76897-912 76897-914
2-Bromo-1,3,4-trichlorobenzene ≥97%
2-Bromo-1,3,4-trichlorobenzene
Formula: C₆H₂BrCl₃
MW: 260.343 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176520
CAS Number: 101654-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR