2-Fluoro-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

20310-1G 20310-25G 20310-500MG 20310-5G
76897-880EA 166.57 USD
76897-880 76897-882 76897-884 76897-886
2-Fluoro-4-isopropoxybenzoic acid ≥97%
2-Fluoro-4-isopropoxybenzoic acid
Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD00142916
CAS Number: 289039-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR