2-Bromo-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-4-fluorophenyl)boronic acid

20149-1G 20149-25G 20149-5G
76897-802EA 111.4 USD
76897-802 76897-804 76897-806
2-Bromo-4-fluorophenylboronic acid ≥97%
2-Bromo-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195647
CAS Number: 1217501-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR