2-Propoxy-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Propoxypyridine-3-boronic acid, (2-Propoxypyridin-3-yl)boronic acid

20061-1G 20061-25G 20061-5G 20061-500MG
76897-772EA 96.55 USD
76897-772 76897-774 76897-776 77645-132
2-Propoxy-3-pyridineboronic acid ≥97%
2-Propoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
MW: 181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195680
CAS Number: 1218790-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR