2-Bromo-4-fluoro-1-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

19946-1G 19946-25G 19946-5G
76897-742EA 84.88 USD
76897-742 76897-744 76897-746
2-Bromo-4-fluoro-1-propoxybenzene ≥95%
2-Bromo-4-fluoro-1-propoxybenzene
Formula: C₉H₁₀BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD09940143
CAS Number: 1016889-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR