2-Fluoro-5-isobutoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19943-1G 19943-25G 19943-5G
76897-730EA 94.43 USD
76897-730 76897-732 76897-734
2-Fluoro-5-isobutoxyphenylboronic acid ≥97%
2-Fluoro-5-isobutoxyphenylboronic acid
Formula: C₁₀H₁₄BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD12546461
CAS Number: 1217500-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR