2-Bromo-6-chlorophenol ≥97%

Supplier: AOBChem USA

Warning

19932-25G 19932-5G
76897-720EA 304.48 USD
76897-720 76897-722
2-Bromo-6-chlorophenol ≥97%
2-Bromo-6-chlorophenol
Formula: C₆H₄BrClO
Storage Temperature: Ambient
MDL Number: MFCD09907875
CAS Number: 2040-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR