2-Bromo-5-chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-5-chlorophenyl)boronic acid

19906-1G 19906-25G 19906-500MG 19906-5G
76897-698EA 173.99 USD
76897-698 76897-700 76897-702 76897-704
2-Bromo-5-chlorophenylboronic acid ≥97%
2-Bromo-5-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12026715
CAS Number: 1217501-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR