Ethyl 2-bromo-5-(trifluoromethyl)benzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Bromo-5-(trifluoromethyl)benzoic acid ethyl ester
19867-25G
19867-5G
76897-690EA
212.18
USD
76897-690
76897-692
Ethyl 2-bromo-5-(trifluoromethyl)benzoate ≥97%
Ethyl 2-bromo-5-(trifluoromethyl)benzoate
Formula:
C₁₀H₈BrF₃O₂ MW: 297.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09999462 CAS Number: 1214336-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...