2-Bromo-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

19845-25G 19845-5G
76897-678EA 396.78 USD
76897-678 76897-680
2-Bromo-N-methoxy-N-methylbenzamide ≥97%
2-Bromo-N-methoxy-N-methylbenzamide
Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD12798299
CAS Number: 899425-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR