2-Chloro-6-(trifluoromethyl)benzoic acid ethyl ester ≥97% research grade

Supplier: AOBChem USA

Warning

19789-1G 19789-25G 19789-500MG 19789-5G
76897-662EA 379.81 USD
76897-662 76897-664 76897-666 76897-668
2-Chloro-6-(trifluoromethyl)benzoic acid ethyl ester ≥97% research grade
2-Chloro-6-(trifluoromethyl)benzoic acid ethyl ester
Formula: C₁₀H₈ClF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD14698202
CAS Number: 1214346-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR