2-Bromo-4-fluoro-1-(methylsulfonyl)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-fluorophenyl methyl sulphone

19695-1G 19695-25G 19695-500MG 19695-5G
76897-648EA 165.51 USD
76897-648 76897-650 76897-652 76897-654
2-Bromo-4-fluoro-1-(methylsulfonyl)benzene ≥97% research grade
2-Bromo-4-fluoro-1-(methylsulphonyl)benzene
Formula: C₇H₆BrFO₂S
MW: 253.0887 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195736
CAS Number: 1039744-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR