1-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Acetylphenylboronic acid pinacol ester
19662-1G
19662-25G
19662-5G
76897-636EA
64.72
USD
76897-636
76897-638
76897-640
1-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone ≥97% research grade
1-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone
Formula:
C₁₄H₁₉BO₃ Storage Temperature: Ambient |
MDL Number:
MFCD18447600 CAS Number: 325141-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...