2-(2,6-Difluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-3-nitrophenylboronic acid pinacol ester

19645-1G 19645-25G 19645-500MG 19645-5G
76897-620EA 138.98 USD
76897-620 76897-622 76897-624 76897-626
2-(2,6-Difluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2,6-Difluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₂H₁₄BF₂NO₄
MW: 285.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295275
CAS Number: 1451391-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR