2-Chloro-5-isopropylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

19415-1G 19415-25G 19415-500MG 19415-5G
76897-566EA 201.58 USD
76897-566 76897-568 76897-570 76897-572
2-Chloro-5-isopropylbenzoic acid ≥95%
2-Chloro-5-isopropylbenzoic acid
Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845954
CAS Number: 1160575-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR