2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzoic acid
19387-1G
19387-25G
19387-5G
76897-556EA
81.69
USD
76897-556
76897-558
76897-560
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid ≥97%
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Formula:
C₁₃H₁₅BF₂O₄ MW: 284.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16996277 CAS Number: 1008119-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...