(2,4-Difluoro-3-(methoxycarbonyl)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Difluoro-3-methoxycarbonylphenylboronic acid

19164-1G 19164-25G 19164-500MG 19164-5G
76897-494EA 249.32 USD
76897-494 76897-496 76897-498 76897-500
(2,4-Difluoro-3-(methoxycarbonyl)phenyl)boronic acid ≥97% research grade
(2,4-Difluoro-3-(methoxycarbonyl)phenyl)boronic acid
Formula: C₈H₇BF₂O₄
MW: 215.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295260
CAS Number: 1190989-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR