2-Methylthiophene-3-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

19149-1G 19149-25G 19149-500MG 19149-5G
76897-478EA 142.17 USD
76897-478 76897-480 76897-482 76897-484
2-Methylthiophene-3-boronic acid pinacol ester ≥97% research grade
2-Methylthiophene-3-boronic acid pinacol ester
Formula: C₁₁H₁₇BO₂S
Storage Temperature: Ambient
MDL Number: MFCD09837623
CAS Number: 910553-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR