2-Bromo-5-chloro-1,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

19093-25G 19093-5G
76897-470EA 198.39 USD
76897-470 76897-472
2-Bromo-5-chloro-1,3-difluorobenzene ≥97%
2-Bromo-5-chloro-1,3-difluorobenzene
Formula: C₆H₂BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD06798002
CAS Number: 883546-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR