2-(2-Fluoro-3-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-isopropoxyphenylboronic acid pinacol ester

19077-1G 19077-25G 19077-500MG 19077-5G
76897-460EA 153.84 USD
76897-460 76897-462 76897-464 76897-466
2-(2-Fluoro-3-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-Fluoro-3-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₅H₂₂BFO₃
MW: 280.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295258
CAS Number: 1451391-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR