2-Fluoro-5-hydroxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoro-4-methylpyridin-3-ol, 2-Fluoro-5-hydroxy-4-picoline

19066-1G 19066-25G 19066-5G
76897-448EA 106.09 USD
76897-448 76897-450 76897-452
2-Fluoro-5-hydroxy-4-methylpyridine ≥97%
2-Fluoro-5-hydroxy-4-methylpyridine
Formula: C₆H₆FNO
MW: 127.116 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611074
CAS Number: 1227577-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR