2-Fluoro-5-iodo-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

19055-1G 19055-25G 19055-5G
76897-440EA 92.3 USD
76897-440 76897-442 76897-444
2-Fluoro-5-iodo-4-methylpyridine ≥97%
2-Fluoro-5-iodo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12911585
CAS Number: 1184913-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR